3-Chlor-4-iodbenzotrifluorid
Lieferant: CHEMPUR
FL4483.POR
004483-1G
004483-5G
CHMPFL4483.POREA
0
EUR
CHMPFL4483.POR
FLUO004483-1G
FLUO004483-5G
3-Chlor-4-iodbenzotrifluorid
3-Chlor-4-iodbenzotrifluorid
Formel:
C₇H₃ClF₃I Molecular Weight: 306.45 g/mol |
MDL:
MFCD00042562 CAS-Nummer: 141738-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...