3-Chlor-4-iodbenzotrifluorid

Lieferant: CHEMPUR

FL4483.POR 004483-1G 004483-5G
CHMPFL4483.POREA 0 EUR
CHMPFL4483.POR FLUO004483-1G FLUO004483-5G
3-Chlor-4-iodbenzotrifluorid
3-Chlor-4-iodbenzotrifluorid
Formel: C₇H₃ClF₃I
Molecular Weight: 306.45 g/mol
MDL: MFCD00042562
CAS-Nummer: 141738-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR