4-Chlor-3-iodbenzotrifluorid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-5-(trifluormethyl)iodbenzol, 4-Chlor-3-iodbenzotrifluorid, 1-Chlor-2-iod-4-(trifluormethyl)benzol
004484-25G
004484-100G
FLUO004484-25GEA
34.1
EUR
FLUO004484-25G
FLUO004484-100G
4-Chlor-3-iodbenzotrifluorid
4-Chlor-3-iodbenzotrifluorid
Formel:
C₇H₃ClF₃I Molecular Weight: 306.45 g/mol Siedepunkt: 58 Dichte: 1.92 g/cm³ (20 °C) |
MDL:
MFCD00001037 CAS-Nummer: 672-57-1 EINECS: 211-593-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...