4-Chlor-3-iodbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-5-(trifluormethyl)iodbenzol, 4-Chlor-3-iodbenzotrifluorid, 1-Chlor-2-iod-4-(trifluormethyl)benzol

004484-25G 004484-100G
FLUO004484-25GEA 34.1 EUR
FLUO004484-25G FLUO004484-100G
4-Chlor-3-iodbenzotrifluorid
4-Chlor-3-iodbenzotrifluorid
Formel: C₇H₃ClF₃I
Molecular Weight: 306.45 g/mol
Siedepunkt: 58
Dichte: 1.92 g/cm³ (20 °C)
MDL: MFCD00001037
CAS-Nummer: 672-57-1
EINECS: 211-593-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR