Isofluran

Lieferant: FLUOROCHEM

004529-5G
FLUO004529-5GEA 61.1 EUR
FLUO004529-5G
Isofluran
Isofluran
Formel: C₃H₂ClF₅O
MDL: MFCD00066609
CAS-Nummer: 26675-46-7
EINECS: 247-897-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR