4-Chlor-7-(trifluormethyl)chinolin

Lieferant: FLUOROCHEM

Synonyme: 4-Chloro-7-trifluoromethylquinoline

CFF992.1 CFF992.10 CFF992.25 004561-5G 004561-1G
CHMPCFF992.1EA 522 EUR
CHMPCFF992.1 CHMPCFF992.10 CHMPCFF992.25 FLUO004561-5G FLUO004561-1G
4-Chlor-7-(trifluormethyl)chinolin
4-Chloro-7-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
Molecular Weight: 231.6016 g/mol
MDL: MFCD00006775
CAS-Nummer: 346-55-4
EINECS: 206-471-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR