4-Chlor-8-(trifluormethyl)chinolin

Lieferant: FLUOROCHEM

CFF993.1 004562-1G 004562-5G
CHMPCFF993.1EA 522 EUR
CHMPCFF993.1 FLUO004562-1G FLUO004562-5G
4-Chlor-8-(trifluormethyl)chinolin
4-Chloro-8-(trifluoromethyl)quinoline
Formel: C₁₀H₅ClF₃N
MDL: MFCD00134578
CAS-Nummer: 23779-97-7
EINECS: 245-880-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR