2-Nitro-4-(trifluormethyl)benzonitril
Lieferant: FLUOROCHEM
Synonyme:
4-Cyano-3-nitrobenzotrifluoride
CFF1252.1
004592-100G
004592-50G
004592-500G
CHMPCFF1252.1EA
522
EUR
CHMPCFF1252.1
FLUO004592-100G
FLUO004592-50G
FLUO004592-500G
2-Nitro-4-(trifluormethyl)benzonitril
2-Nitro-4-(trifluormethyl)benzonitril
Formel:
C₈H₃F₃N₂O₂ |
MDL:
MFCD00014684 CAS-Nummer: 778-94-9 EINECS: 212-298-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...