2-Nitro-4-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

Synonyme: 4-Cyano-3-nitrobenzotrifluoride

CFF1252.1 004592-100G 004592-50G 004592-500G
CHMPCFF1252.1EA 522 EUR
CHMPCFF1252.1 FLUO004592-100G FLUO004592-50G FLUO004592-500G
2-Nitro-4-(trifluormethyl)benzonitril
2-Nitro-4-(trifluormethyl)benzonitril
Formel: C₈H₃F₃N₂O₂
MDL: MFCD00014684
CAS-Nummer: 778-94-9
EINECS: 212-298-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR