4'-Cyan-2'-(trifluormethoxy)acetanilid
Lieferant: FLUOROCHEM
004593-1G
FLUO004593-1GEA
39.9
EUR
FLUO004593-1G
4'-Cyan-2'-(trifluormethoxy)acetanilid
4'-Cyan-2'-(trifluormethoxy)acetanilid
Formel:
C₁₀H₇F₃N₂O₂ |
MDL:
MFCD00204174 CAS-Nummer: 175278-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...