4'-Cyan-2'-(trifluormethyl)acetanilid

Lieferant: FLUOROCHEM

004594-1G 004594-5G
FLUO004594-1GEA 88.1 EUR
FLUO004594-1G FLUO004594-5G
4'-Cyan-2'-(trifluormethyl)acetanilid
4'-Cyan-2'-(trifluormethyl)acetanilid
Formel: C₁₀H₇F₃N₂O
MDL: MFCD00052844
CAS-Nummer: 175277-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR