4-(Trifluormethyl)nicotinonitril
Lieferant: FLUOROCHEM
004597-1G
FLUO004597-1GEA
130
EUR
FLUO004597-1G
4-(Trifluormethyl)nicotinonitril
4-(Trifluormethyl)nicotinonitril
Formel:
C₇H₃F₃N₂ |
MDL:
MFCD00153107 CAS-Nummer: 13600-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...