2,6-Dibrom-4-fluoranilin
Lieferant: FLUOROCHEM
004650-25G
004650-100G
004650-500G
FLUO004650-25GEA
36.9
EUR
FLUO004650-25G
FLUO004650-100G
FLUO004650-500G
2,6-Dibrom-4-fluoranilin
2,6-Dibrom-4-fluoranilin
Formel:
C₆H₄Br₂FN Siedepunkt: 277 °C (1013 hPa) Schmelzpunkt: 68 °C |
MDL:
MFCD00041445 CAS-Nummer: 344-18-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...