2,4-Dibrom-1-fluorbenzol

Lieferant: FLUOROCHEM

Synonyme: 1,3-Dibromo-4-fluorobenzene, 1-Fluoro-2,4-dibromobenzene

004653-25G
FLUO004653-25GEA 48.6 EUR
FLUO004653-25G
2,4-Dibrom-1-fluorbenzol
2,4-Dibrom-1-fluorbenzol
Formel: C₆H₃Br₂F
Molecular Weight: 253,9 g/mol
MDL: MFCD00000283
CAS-Nummer: 1435-53-6
EINECS: 215-860-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR