1,3-Dibromtetrafluorbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
004667-25G
004667-5G
FLUO004667-25GEA
0
EUR
FLUO004667-25G
FLUO004667-5G
1,3-Dibromtetrafluorbenzol
1,3-Dibromtetrafluorbenzol
Formel:
C₆Br₂F₄ Siedepunkt: 85 °C (18 mmHg) |
MDL:
MFCD00031533 CAS-Nummer: 1559-87-1 EINECS: 216-326-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...