2,6-Dibrom-4-(trifluormethoxy)anilin

Lieferant: FLUOROCHEM

004677-1G 004677-5G 004677-25G 004677-100G
FLUO004677-1GEA 30.3 EUR
FLUO004677-1G FLUO004677-5G FLUO004677-25G FLUO004677-100G
2,6-Dibrom-4-(trifluormethoxy)anilin
2,6-Dibrom-4-(trifluormethoxy)anilin
Formel: C₇H₄Br₂F₃NO
MDL: MFCD00153113
CAS-Nummer: 88149-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR