2,4-Dichlor-3,5-dinitro-α,α,α-trifluortoluol
Lieferant: FLUOROCHEM
Synonyme:
1,3-Dichloro-2,4-dinitro-6-(trifluoromethyl)benzene, 2,4-Dichloro-3,5-dinitrobenzotrifluoride, 2,4-Dichloro-1,3-dinitro-5-(trifluoromethyl)benzene
004703-5G
004703.25G
FLUO004703-5GEA
0
EUR
FLUO004703-5G
FLUO004703.25G
2,4-Dichlor-3,5-dinitro-α,α,α-trifluortoluol
2,4-Dichlor-3,5-dinitro-α,α,α-trifluortoluol
Formel:
C₇HCl₂F₃N₂O₄ Molecular Weight: 305 g/mol Siedepunkt: 291…294 °C (1013 hPa) Schmelzpunkt: 74…75 °C |
MDL:
MFCD00042480 CAS-Nummer: 29091-09-6 EINECS: 249-420-8 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...