1,2-Dichlor-1,1,2-trifluor-2-iodethan

Lieferant: FLUOROCHEM

004737-25G
FLUO004737-25GEA 0 EUR
FLUO004737-25G
1,2-Dichlor-1,1,2-trifluor-2-iodethan
1,2-Dichlor-1,1,2-trifluor-2-iodethan
Formel: ClF₂CCClFI
MDL: MFCD00039402
CAS-Nummer: 354-61-0
EINECS: 206-565-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR