2,4-Dichlor-6-(trifluormethyl)anilin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Amino-3,5-dichlor-α,α,α-trifluortoluol, 4,6-Dichlor-α,α,α-trifluor-o-toluidin

004740-1G
FLUO004740-1GEA 37.2 EUR
FLUO004740-1G
2,4-Dichlor-6-(trifluormethyl)anilin
2,4-Dichlor-6-(trifluormethyl)anilin
Formel: C₇H₄Cl₂F₃N
Molecular Weight: 230,02 g/mol
MDL: MFCD00174313
CAS-Nummer: 62593-17-3
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR