2',4'-Difluoracetanilid
Lieferant: FLUOROCHEM
Synonyme:
N-(2,4-Difluorphenyl)acetamid
004774-100G
FLUO004774-100GEA
72.9
EUR
FLUO004774-100G
2',4'-Difluoracetanilid
2',4'-Difluoracetanilid
Formel:
C₈H₇F₂NO |
MDL:
MFCD00032502 CAS-Nummer: 399-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...