2,3-Difluorbenzophenon
Lieferant: FLUOROCHEM
004783-5G
FLUO004783-5GEA
0
EUR
FLUO004783-5G
2,3-Difluorbenzophenon
2,3-Difluorbenzophenon
Formel:
C₁₃H₈F₂O Molecular Weight: 218.2 g/mol |
MDL:
MFCD00061139 CAS-Nummer: 208173-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...