(±)-3,5-Difluormandelsäure

Lieferant: FLUOROCHEM

Synonyme: (±)-3,5-Difluorphenylglykolsäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyethansäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyessigsäure

004826-1G 004826-5G 004826-25G
FLUO004826-1GEA 31.7 EUR
FLUO004826-1G FLUO004826-5G FLUO004826-25G
(±)-3,5-Difluormandelsäure
(±)-3,5-Difluormandelsäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188.13 g/mol
MDL: MFCD00061300
CAS-Nummer: 132741-31-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR