(±)-3,5-Difluormandelsäure
Lieferant: FLUOROCHEM
Synonyme:
(±)-3,5-Difluorphenylglykolsäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyethansäure, (±)-2-(3,5-Difluorphenyl)-2-hydroxyessigsäure
004826-1G
004826-5G
004826-25G
FLUO004826-1GEA
31.7
EUR
FLUO004826-1G
FLUO004826-5G
FLUO004826-25G
(±)-3,5-Difluormandelsäure
(±)-3,5-Difluormandelsäure
Formel:
C₈H₆F₂O₃ Molecular Weight: 188.13 g/mol |
MDL:
MFCD00061300 CAS-Nummer: 132741-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...