4-(Difluormethoxy)anilin
Lieferant: FLUOROCHEM
|
Achtung
|
004828-5G
004828-25G
FLUO004828-5GEA
36.9
EUR
FLUO004828-5G
FLUO004828-25G
4-(Difluormethoxy)anilin
4-(Difluormethoxy)anilin
Formel:
C₇H₇F₂NO Siedepunkt: 90 °C (1013 hPa) Dichte: 1,286 g/cm³ (20 °C) |
MDL:
MFCD00085005 CAS-Nummer: 22236-10-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...