1-Brom-4-(difluormethoxy)benzol
Lieferant: FLUOROCHEM
Synonyme:
4-(Difluoromethoxy)bromobenzene
004836-1G
004836.5G
004836.25G
FLUO004836-1GEA
32.4
EUR
FLUO004836-1G
FLUO004836.5G
FLUO004836.25G
1-Brom-4-(difluormethoxy)benzol
1-Brom-4-(difluormethoxy)benzol
Formel:
C₇H₅BrF₂O Molecular Weight: 223,02 g/mol |
MDL:
MFCD00221465 CAS-Nummer: 5905-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...