2,4-Difluor-6-nitroanilin
Lieferant: FLUOROCHEM
CFF1043.1
CFF1043.10
004845-25G
004845-10G
004845-100G
CHMPCFF1043.1EA
522
EUR
CHMPCFF1043.1
CHMPCFF1043.10
FLUO004845-25G
FLUO004845-10G
FLUO004845-100G
2,4-Difluor-6-nitroanilin
2,4-Difluor-6-nitroanilin
Formel:
C₆H₄F₂N₂O₂ Molecular Weight: 174.11 g/mol |
MDL:
MFCD00034058 CAS-Nummer: 364-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...