2,4-Difluor-6-nitroanilin

Lieferant: FLUOROCHEM

CFF1043.1 CFF1043.10 004845-25G 004845-10G 004845-100G
CHMPCFF1043.1EA 522 EUR
CHMPCFF1043.1 CHMPCFF1043.10 FLUO004845-25G FLUO004845-10G FLUO004845-100G
2,4-Difluor-6-nitroanilin
2,4-Difluor-6-nitroanilin
Formel: C₆H₄F₂N₂O₂
Molecular Weight: 174.11 g/mol
MDL: MFCD00034058
CAS-Nummer: 364-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR