2,4-Difluor-DL-phenylalanin

Lieferant: FLUOROCHEM

004856-1G
FLUO004856-1GEA 159 EUR
FLUO004856-1G
2,4-Difluor-DL-phenylalanin
2,4-Difluor-DL-phenylalanin
Formel: C₉H₉F₂NO₂
MDL: MFCD00061302
CAS-Nummer: 32133-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR