3,4-Difluor-DL-phenylalanin

Lieferant: FLUOROCHEM

004858-1G
FLUO004858-1GEA 164 EUR
FLUO004858-1G
3,4-Difluor-DL-phenylalanin
3,4-Difluor-DL-phenylalanin
Formel: C₉H₉F₂NO₂
Molecular Weight: 201,18 g/mol
MDL: MFCD00061307
CAS-Nummer: 32133-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR