4-Fluorveratrol

Lieferant: FLUOROCHEM

004913-5G
FLUO004913-5GEA 46.2 EUR
FLUO004913-5G
4-Fluorveratrol
4-Fluorveratrol
Formel: C₈H₉FO₂
MDL: MFCD00012201
CAS-Nummer: 398-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR