2',4'-Bis(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-[2,4-Bis(trifluormethyl)phenyl]ethan-1-on

004995-1G 004995.5G
FLUO004995-1GEA 114 EUR
FLUO004995-1G FLUO004995.5G
2',4'-Bis(trifluormethyl)acetophenon
2',4'-Bis(trifluormethyl)acetophenon
Formel: C₁₀H₆F₆O
Molecular Weight: 256,15 g/mol
MDL: MFCD00042495
CAS-Nummer: 237069-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR