3-Chlor-5-fluorbenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
CFF934.1
005181-1G
005181-25G
005181-5G
CHMPCFF934.1EA
522
EUR
CHMPCFF934.1
FLUO005181-1G
FLUO005181-25G
FLUO005181-5G
3-Chlor-5-fluorbenzoesäure
3-Chloro-5-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174.56 g/mol |
MDL:
MFCD00973904 CAS-Nummer: 25026-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...