3-Chlor-5-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

CFF934.1 005181-1G 005181-25G 005181-5G
CHMPCFF934.1EA 522 EUR
CHMPCFF934.1 FLUO005181-1G FLUO005181-25G FLUO005181-5G
3-Chlor-5-fluorbenzoesäure
3-Chloro-5-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174.56 g/mol
MDL: MFCD00973904
CAS-Nummer: 25026-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR