5-Fluor-2-mercaptobenzothiazol

Lieferant: FLUOROCHEM

Synonyme: 5-Fluorobenzo[d]thiazole-2-thiol

005187-1G
FLUO005187-1GEA 282 EUR
FLUO005187-1G
5-Fluor-2-mercaptobenzothiazol
5-Fluoro-2-mercaptobenzothiazole
Formel: C₇H₄FNS₂
Molecular Weight: 185,25 g/mol
MDL: MFCD02181177
CAS-Nummer: 155559-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR