5-Brom-2-hydroxybenzonitril

Lieferant: FLUOROCHEM

CFF852.1 005188-25G 005188-5G
CHMPCFF852.1EA 522 EUR
CHMPCFF852.1 FLUO005188-25G FLUO005188-5G
5-Brom-2-hydroxybenzonitril
5-Brom-2-hydroxybenzonitril
Formel: C₇H₄BrNO
MDL: MFCD00143428
CAS-Nummer: 40530-18-5
EINECS: 254-958-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR