5-Brom-2-thiophensulfonylchlorid

Lieferant: FLUOROCHEM

CFF581.10 005201-5G 005201-25G 005201-100G
CHMPCFF581.10EA 348 EUR
CHMPCFF581.10 FLUO005201-5G FLUO005201-25G FLUO005201-100G
5-Brom-2-thiophensulfonylchlorid
5-Brom-2-thiophensulfonylchlorid
Formel: C₄H₂BrClO₂S₂
MDL: MFCD00084909
CAS-Nummer: 55854-46-1
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR