4-Brom-2-fluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

005240-25G
FLUO005240-25GEA 87 EUR
FLUO005240-25G
4-Brom-2-fluorbenzolsulfonylchlorid
4-Brom-2-fluorbenzolsulfonylchlorid
Formel: C₆H₃BrClFO₂S
Molecular Weight: 273,51 g/mol
Siedepunkt: 84 °C (0,5 mmHg)
Schmelzpunkt: 63…66 °C
MDL: MFCD01090993
CAS-Nummer: 216159-03-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR