2-Fluor-5-iodtoluol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-Fluoro-4-iodo-2-methylbenzene, 2-Fluoro-5-iodo-1-methylbenzene

CFF1116. 005313-1G 005313-5G 005313-25G
CHMPCFF1116.EA 0 EUR
CHMPCFF1116. FLUO005313-1G FLUO005313-5G FLUO005313-25G
2-Fluor-5-iodtoluol
2-Fluor-5-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236.03 g/mol
MDL: MFCD00013710
CAS-Nummer: 452-68-6
EINECS: 207-206-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR