1-(4-Fluorphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 4-Fluorphenylthioharnstoff, N-(4-Fluorphenyl)thioharnstoff

005417-5G 005417-25G 005417-100G
FLUO005417-5GEA 34.1 EUR
FLUO005417-5G FLUO005417-25G FLUO005417-100G
1-(4-Fluorphenyl)-2-thioharnstoff
1-(4-Fluorphenyl)-2-thioharnstoff
Formel: (C₆H₄F)NHC(S)NH₂
Molecular Weight: 170.21 g/mol
Siedepunkt: 276
Schmelzpunkt: 169…170 °C
MDL: MFCD00041180
CAS-Nummer: 459-05-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR