4-Chlor-2-iodanilin
Lieferant: FLUOROCHEM
005435-25G
005435-100G
FLUO005435-25GEA
63.9
EUR
FLUO005435-25G
FLUO005435-100G
4-Chlor-2-iodanilin
4-Chlor-2-iodanilin
Formel:
C₆H₅ClIN Molecular Weight: 253.47 g/mol |
MDL:
MFCD01863737 CAS-Nummer: 63069-48-7 EINECS: 263-839-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...