2-(4-Aminophenyl)ethanol

Lieferant: FLUOROCHEM

Synonyme: 4-Aminophenethylalkohol, 2-(4-Aminophenyl)ethylalkohol

005442-25G
FLUO005442-25GEA 69.6 EUR
FLUO005442-25G
2-(4-Aminophenyl)ethanol
2-(4-Aminophenyl)ethanol
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Siedepunkt: 235…255 °C (1013 hPa)
Schmelzpunkt: 108 °C
MDL: MFCD00007922
CAS-Nummer: 104-10-9
EINECS: 203-174-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR