4-Iod-o-toluidin
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-5-iodtoluol, 4-Iod-2-methylanilin
CFF1189.
005444-5G
005444-25G
CHMPCFF1189.EA
0
EUR
CHMPCFF1189.
FLUO005444-5G
FLUO005444-25G
4-Iod-o-toluidin
4-Iod-o-toluidin
Formel:
C₇H₈IN Molecular Weight: 233.05 g/mol Siedepunkt: 279 Schmelzpunkt: 91…92 °C |
MDL:
MFCD00025299 CAS-Nummer: 13194-68-8 EINECS: 236-154-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...