2-Brom-3-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

CFF837.1 005469-1G
CHMPCFF837.1EA 522 EUR
CHMPCFF837.1 FLUO005469-1G
2-Brom-3-fluorbenzoesäure
2-Bromo-3-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD01569398
CAS-Nummer: 132715-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR