3-Brom-5-fluorbenzoesäure

Lieferant: CHEMPUR

CFF843.1 FL5475.500 005475-5G 005475-1G
CHMPCFF843.1EA 522 EUR
CHMPCFF843.1 CHMPFL5475.500 FLUO005475-5G FLUO005475-1G
3-Brom-5-fluorbenzoesäure
3-Bromo-5-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219.01 g/mol
MDL: MFCD01569540
CAS-Nummer: 176548-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR