4-Amino-6-(trifluormethyl)chinolin

Lieferant: FLUOROCHEM

CFF812.1 005478-500MG 005478-1G
CHMPCFF812.1EA 522 EUR
CHMPCFF812.1 FLUO005478-500MG FLUO005478-1G
4-Amino-6-(trifluormethyl)chinolin
4-Amino-6-(trifluoromethyl)quinoline
Formel: C₁₀H₇F₃N₂
MDL: MFCD00278781
CAS-Nummer: 247113-89-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR