4-Amino-7-(trifluormethyl)chinolin

Lieferant: FLUOROCHEM

CFF813.1 005479-500MG 005479-1G
CHMPCFF813.1EA 522 EUR
CHMPCFF813.1 FLUO005479-500MG FLUO005479-1G
4-Amino-7-(trifluormethyl)chinolin
4-Amino-7-(trifluoromethyl)quinoline
Formel: C₁₀H₇F₃N₂
Molecular Weight: 212.18 g/mol
MDL: MFCD00269411
CAS-Nummer: 243666-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR