6-Fluor-4-chinolinol

Lieferant: CHEMPUR

Synonyme: 6-Fluoro-4-hydroxyquinoline

CFF1103.1 FL5483.500 005483-1G 005483-5G
CHMPCFF1103.1EA 522 EUR
CHMPCFF1103.1 CHMPFL5483.500 FLUO005483-1G FLUO005483-5G
6-Fluor-4-chinolinol
6-Fluor-4-chinolinol
Formel: C₉H₆FNO
Molecular Weight: 163.1485 g/mol
MDL: MFCD00269610
CAS-Nummer: 391-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR