8-Fluor-4-chinolinol
Lieferant: FLUOROCHEM
Synonyme:
8-Fluoroquinolin-4-ol, 8-Fluoro-4-hydroxyquinoline
CFF1105.1
005484-1G
CHMPCFF1105.1EA
522
EUR
CHMPCFF1105.1
FLUO005484-1G
8-Fluor-4-chinolinol
8-Fluor-4-chinolinol
Formel:
C₉H₆FNO Molecular Weight: 163,15 g/mol |
MDL:
MFCD00272395 CAS-Nummer: 63010-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...