4-Fluor-3-methoxybenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 3-Methoxy-4-fluorobenzoic acid

CFF1125. 005490-1G
CHMPCFF1125.EA 0 EUR
CHMPCFF1125. FLUO005490-1G
4-Fluor-3-methoxybenzoesäure
4-Fluoro-3-methoxybenzoic acid
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
MDL: MFCD00272134
CAS-Nummer: 82846-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR