5-Amino-4-brom-1-methyl-1H-pyrazol
Lieferant: FLUOROCHEM
Synonyme:
4-Bromo-1-methyl-1H-pyrazol-5-amine
CFF537.1
005503-1G
CHMPCFF537.1EA
508
EUR
CHMPCFF537.1
FLUO005503-1G
5-Amino-4-brom-1-methyl-1H-pyrazol
5-Amino-4-bromo-1-methyl-1H-pyrazole
Formel:
C₄H₆BrN₃ Molecular Weight: 176,02 g/mol |
MDL:
MFCD00068157 CAS-Nummer: 105675-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...